Anti-IDH1 Goat Polyclonal Antibody

Supplier: ProSci
42-705
10113-650EA 701.94 USD
10113-650
Anti-IDH1 Goat Polyclonal Antibody
Antibodies
Approximately 48 kDa band observed in HEPG2 lysate (calculated MW of 46.7 kDa according to NP_005887.2). Recommended concentration: 0.3-1 ?g/mL.

IDH1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR