Anti-EIF3E Goat Polyclonal Antibody

Supplier: ProSci
42-702
10114-780EA 701.94 USD
10114-780
Anti-EIF3E Goat Polyclonal Antibody
Antibodies
Approximately 50 kDa band observed in lysates of Daudi lysate (calculated MW of 52.2 kDa according to NP_001559.1). Recommended concentration: 0.03-0.1 ?g/mL.

EIF3E antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR