Anti-TGM7 Goat Polyclonal Antibody

Supplier: ProSci
42-698
10114-776EA 701.94 USD
10114-776
Anti-TGM7 Goat Polyclonal Antibody
Antibodies
Approximately 75 kDa band observed in human lung and testis and in mouse kidney, lung and testis lysates (calculated MW of 79.9 kDa according to human NP_443187.1 and 79.8 kDa according to mouse NP_001153896.1). Recommended concentration: 0.1-0.3 ?g/mL.
an additional band of unknown identity was also consistently observed at 17 kDa . This band was successfully blocked by incubation with the immunizing peptide.

TGM7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TGM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR