Anti-TH1L Goat Polyclonal Antibody

Supplier: ProSci
42-694
10114-772EA 701.94 USD
10114-772
Anti-TH1L Goat Polyclonal Antibody
Antibodies
Approximately 60 kDa band observed in nuclear Jurkat lysate (calculated MW of 66.2 kDa according to NP_945327.1). Recommended concentration: 0.5-2 ?g/mL.

TH1L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TH1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR