Anti-DCTN3 Goat Polyclonal Antibody

Supplier: ProSci
42-682
10114-760EA 701.94 USD
10114-760
Anti-DCTN3 Goat Polyclonal Antibody
Antibodies
Approximately 20 kDa band observed in human kidney, skeletal muscle and pancreas lysates (calculated MW of 19.5 kDa according to NP_077324.1). Recommended concentration: 0.03-0.1 ?g/mL.

DCTN3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DCTN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR