Anti-ALDH6A1 Goat Polyclonal Antibody

Supplier: ProSci

42-679
10113-630EA 560.36 USD
10113-630
Anti-ALDH6A1 Goat Polyclonal Antibody
Antibodies
Approximately 55 kDa band observed in human, mouse and rat kidney lysates (calculated MW of 57.8 kDa according to NP_005580.1). Recommended concentration: 0.1-0.3 ?g/mL.

ALDH6A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALDH6A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR