Anti-ALDH5A1 Goat Polyclonal Antibody

Supplier: ProSci
42-676
10113-624EA 701.94 USD
10113-624
Anti-ALDH5A1 Goat Polyclonal Antibody
Antibodies
Approximately 55 kDa band observed in human liver lysates (calculated MW of 58.7 kDa according to NP_733936.1). Recommended concentration: 0.3-1 ?g/mL. Please note that both this antibody and EB10436 detects a 50 kDa in mouse and rat liver lysates (calculated MW of 56.0 kDa according to mouse NP_766120.1).

ALDH5A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALDH5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR