Anti-ALDH18A1 Goat Polyclonal Antibody

Supplier: ProSci
42-675
10114-754EA 701.94 USD
10114-754
Anti-ALDH18A1 Goat Polyclonal Antibody
Antibodies
Approximately 85 kDa band observed in MCF7 lysate (calculated MW of 87.3 kDa according to NP_002851.2). Recommended concentration: 1-3 ?g/mL.

ALDH18A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALDH18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR