Anti-ZDHHC1 Goat Polyclonal Antibody

Supplier: ProSci
42-670
10113-616EA 552.17 USD
10113-616
Anti-ZDHHC1 Goat Polyclonal Antibody
Antibodies
Approximately 48 kDa and 20 kDa in HEK293 lysate after 2 ?g/mL antibody staining.8 kDa according to NP_037436.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands).

ZDHHC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZDHHC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR