Anti-BCL2A1 Goat Polyclonal Antibody

Supplier: ProSci
42-665
10114-746EA 701.94 USD
10114-746
Anti-BCL2A1 Goat Polyclonal Antibody
Antibodies
Approximately 20 kDa band observed in human bone marrow lysates (calculated MW of 20.1 kDa according to NP_004040.1). Recommended concentration: 0.1-0.3 ?g/mL.

BCL2A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BCL2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR