Anti-POR Goat Polyclonal Antibody

Supplier: ProSci

42-651
10114-728EA 560.36 USD
10114-728
Anti-POR Goat Polyclonal Antibody
Antibodies
Approximately 90 kDa band observed in human, mouse and rat liver lysates (calculated MW of 77.0 kDa according to NP_000932.3). Recommended concentration: 0.1-0.3 ?g/mL.

POR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR