Anti-MGAT1 Goat Polyclonal Antibody

Supplier: ProSci
42-645
10114-718EA 701.94 USD
10114-718
Anti-MGAT1 Goat Polyclonal Antibody
Antibodies
In transfected HEK293 transiently expressing full-length human MGAT1 (myc and DYKDDDDK tagged), a band of approximately 55 kDa was observed. No bands were observed in mock-transfected HEK293 and the same band was observed using anti-myc tag antibody. Recommended concentration, 0.2-1 ?g/mL.

MGAT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MGAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR