Anti-MID1IP1 Goat Polyclonal Antibody

Supplier: ProSci

42-644
10114-716EA 560.36 USD
10114-716
Anti-MID1IP1 Goat Polyclonal Antibody
Antibodies
In transfected HEK293 transiently expressing full-length human MID1IP1 (myc and DYKDDDDK tagged), a band of approximately 28 kDa was observed. No bands were
observed in mock-transfected HEK293 and the same band was observed using anti-myc tag antibody. Recommended concentration, 0.2-1 ?g/mL.

MID1IP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MID1IP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR