Anti-ARTN Goat Polyclonal Antibody

Supplier: ProSci

42-639
10114-714EA 560.21 USD
10114-714
Anti-ARTN Goat Polyclonal Antibody
Antibodies
Approximately 28 kDa band observed in lysates of cell lines HEK293, HepG2 and Kelly (calculated MW of 22.9 kDa according to NP_003967.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Pandey et al, Endocrinology. 2010 Mar;151 (3) :909-20; PMID: 20118197). Recommended concentration: 0.1-0.3 ?g/mL.

ARTN antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR