Anti-CSF1R Goat Polyclonal Antibody

Supplier: ProSci
42-637
10113-592EA 701.94 USD
10113-592
Anti-CSF1R Goat Polyclonal Antibody
Antibodies
In transfected HEK293 transiently expressing full-length human CSF1R (myc and DYKDDDDK tagged), a band of approximately 200 kDa was observed. No bands were observed in mock-transfected HEK293 and the same band was observed using anti-myc tag antibody. Recommended concentration, 0.1-0.5 ?g/mL.

CSF1R antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CSF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR