Anti-CIRBP Goat Polyclonal Antibody

Supplier: ProSci
42-633
10113-590EA 701.94 USD
10113-590
Anti-CIRBP Goat Polyclonal Antibody
Antibodies
Approximately 18 kDa band observed in mouse and rat testis lysates (calculated MW of 18.6 kDa according to human NP_001271.1 and mouse NP_031731.1). Recommended concentration: 0.01-0.03 ?g/mL.

CIRBP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CIRBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR