Anti-NDUFS1 Goat Polyclonal Antibody

Supplier: ProSci
42-624
10114-696EA 701.94 USD
10114-696
Anti-NDUFS1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000 Western Blot: Approximately 80 kDa band observed in human heart and mouse heart lysates (calculated MW of 79.5 kDa according to NP_004997.4). Recommended concentration: 0.3-1 ?g/mL.

NDUFS1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NDUFS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR