Anti-Aldh3a2 Goat Polyclonal Antibody

Supplier: ProSci

42-616
10114-688EA 560.36 USD
10114-688
Anti-Aldh3a2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000 Western Blot: Approximately 55 kDa band observed in mouse and rat liver lysates (calculated MW of 54.0 kDa according to mouse NP_031463.2 and 54.1 kDa according to rat NP_113919.2). Recommended concentration: 0.03-0.1 ?g/mL.

Aldh3a2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Aldh3a2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR