Anti-GNAS Goat Polyclonal Antibody

Supplier: ProSci
42-594
10114-656EA 701.94 USD
10114-656
Anti-GNAS Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000 Western Blot: Approximately 45 kDa band observed in Jurkat lysate (calculated MW of 45.7 kDa according to NP_000507.1). Recommended concentration: 1-3 ?g/mL.

GNAS antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GNAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR