Anti-POU5F1B Goat Polyclonal Antibody

Supplier: ProSci
42-581
10114-644EA 552.17 USD
10114-644
Anti-POU5F1B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000 Western Blot: Approximately 28 kDa band in human heart lysates after 1 ?g/mL antibody staining.6 kDa according to NP_001153014.1. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

POU5F1B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POU5F1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR