Anti-POU5F1 Goat Polyclonal Antibody

Supplier: ProSci
42-580
10114-642EA 701.94 USD
10114-642
Anti-POU5F1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 40 kDa band observed in lysates of cell line NIH3T3 (calculated MW of 38.2 kDa according to NP_038661.2). Recommended concentration: 1-3 ?g/mL.

POU5F1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POU5F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR