Anti-POU2F1 Goat Polyclonal Antibody

Supplier: ProSci

42-575
10114-632EA 560.36 USD
10114-632
Anti-POU2F1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 100 kDa band observed in nuclear HeLa lysate (calculated MW of 76.5 kDa according to NP_002688.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.5-2 ?g/mL.

POU2F1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POU2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR