Anti-FANCM Goat Polyclonal Antibody

Supplier: ProSci
42-572
10114-626EA 550.88 USD
10114-626
Anti-FANCM Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Preliminary experiments in human amygdala and testis lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

FANCM antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FANCM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR