Anti-TTC8 Goat Polyclonal Antibody

Supplier: ProSci
42-571
10114-624EA 701.94 USD
10114-624
Anti-TTC8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000 Western Blot: Approximately 60 kDa band observed in human testis lysates (calculated MW of 58.4 kDa according to NP_653197.2;). Recommended concentration: 0.2-0.6 ?g/mL.

TTC8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TTC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR