Anti-SLC4A8 Goat Polyclonal Antibody

Supplier: ProSci

42-553
10114-608EA 455.18 USD
10114-608
Anti-SLC4A8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Preliminary experiments in human cerebellum, mouse brain and rat brain lysates, and in lysates of human breast tumor, human kidney and human lymph node gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

SLC4A8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC4A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR