Anti-MECOM Goat Polyclonal Antibody

Supplier: ProSci
42-544
10114-594EA 701.94 USD
10114-594
Anti-MECOM Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000 Western Blot: Approximately 170 kDa band observed in A549 and HepG2 lysate (calculated MW of 126 kDa according to NP_001098547.3). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 1-3 ?g/mL.

MECOM antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MECOM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR