Anti-SIT1 Goat Polyclonal Antibody

Supplier: ProSci
42-535
10114-584EA 552.17 USD
10114-584
Anti-SIT1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000 Western Blot: Preliminary experiments in human lymph node, spleen, tonsil and thymus lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

SIT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR