Anti-LSP1 Goat Polyclonal Antibody

Supplier: ProSci
42-530
10113-532EA 701.94 USD
10113-532
Anti-LSP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 50 kDa band observed in human lymph node, tonsil and peripheral blood mononucleocytes lysates (calculated MW of 37.2 kDa according to NP_002330.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Liu et al, J Exp Med. 2005 Feb 7;201 (3) :409-18; PMID: 15684321). Recommended concentration: 0.05-0.2 ?g/mL.

LSP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR