Anti-MON1A Goat Polyclonal Antibody

Supplier: ProSci
42-514
10113-512EA 701.94 USD
10113-512
Anti-MON1A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 50 kDa band observed in Jurkat lysate (calculated MW of 55.5 kDa according to NP_001135973.1). Recommended concentration: 1-3 ?g/mL.

MON1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MON1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR