Anti-MAPRE3 Goat Polyclonal Antibody

Supplier: ProSci
42-510
10114-558EA 701.94 USD
10114-558
Anti-MAPRE3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 33 kDa band observed in human cerebellum and mouse brain lysates (calculated MW of 32.0 kDa according to human NP_036458.2 and mouse NP_579928.1). Recommended concentration: 0.05-0.2 ?g/mL.

MAPRE3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAPRE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR