Anti-TRIM29 Goat Polyclonal Antibody

Supplier: ProSci
42-505
10114-550EA 701.94 USD
10114-550
Anti-TRIM29 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000 Western Blot: Approximately 70 kDa band observed in human peripheral blood mononucleocytes lysates (calculated MW of 65.8 kDa according to NP_036233.2). Recommended concentration: 0.3-1 ?g/mL.
an additional band of unknown identity was also consistently observed at 37 kDa . This band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human skin shows staining of subnuclear structures and cytoplasm in keratinocytes. Recommended concentration, 1-2 ?g/mL.

TRIM29 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRIM29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR