Anti-PDE1A Goat Polyclonal Antibody

Supplier: ProSci
42-501
10114-542EA 701.94 USD
10114-542
Anti-PDE1A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000 Western Blot: Approximately 55 kDa band observed in human cerebellum lysates (calculated MW of 62.3 kDa according to NP_005010.2). Recommended concentration: 1-3 ?g/mL.

PDE1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDE1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR