Anti-SLC12A2 Goat Polyclonal Antibody

Supplier: ProSci
42-498
10114-536EA 701.94 USD
10114-536
Anti-SLC12A2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000 Western Blot: Approximately 75 kDa band in human, mouse and rat skeletal muscle, heart and brain lysates after 1 ?g/mL antibody staining.1. The 75 kDa band was successfully blocked by incubation with the immunizing peptide.

SLC12A2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC12A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR