Anti-ACAT1 Goat Polyclonal Antibody

Supplier: ProSci
42-494
10114-528EA 701.94 USD
10114-528
Anti-ACAT1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000 Western Blot: Approximately 45 kDa band observed in human liver lysates (calculated MW of 45.2 kDa according to NP_000010.1). Recommended concentration: 0.01-0.03 ?g/mL.

ACAT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR