Anti-SFRP1 Goat Polyclonal Antibody

Supplier: ProSci
42-478
10114-502EA 701.94 USD
10114-502
Anti-SFRP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Imunohistochemistry: In paraffin embedded human kidney shows vesicular staining in epithelial cells of PCT. Recommended concentration, 3-6 ?g/mL.

SFRP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SFRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR