Anti-Lgals1 Goat Polyclonal Antibody

Supplier: ProSci

42-454
10113-496EA 560.36 USD
10113-496
Anti-Lgals1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000 Western Blot: Approximately 15 kDa band observed in rat pancreas lysates (calculated MW of 14.9 kDa according to mouse NP_032521.1 and to rat NP_063969.1). Recommended concentration: 0.5-2 ?g/mL.

Lgals1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Lgals1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR