Anti-Itga11 Goat Polyclonal Antibody

Supplier: ProSci
42-436
10114-472EA 701.94 USD
10114-472
Anti-Itga11 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000 Western Blot: Approximately 140 kDa band observed in mouse skeletal muscle lysates (calculated MW of 133 kDa according to NP_795896.4). Recommended concentration: 1-3 ?g/mL.

Itga11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Itga11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR