Anti-ZEB2 Goat Polyclonal Antibody

Supplier: ProSci
42-432
10114-464EA 701.94 USD
10114-464
Anti-ZEB2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000 Western Blot: Approximately 100+38 kDa bands observed in human liver lysates (calculated MW of 136 kDa according to NP_055610.1). Recommended concentration: 0.2-0.5 ?g/mL. We believe the two observed bands represent two breakdown fragments.

ZEB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR