Anti-Ugt2b Goat Polyclonal Antibody

Supplier: ProSci
42-431
10114-462EA 701.94 USD
10114-462
Anti-Ugt2b Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000 Western Blot: Approximately 50 kDa band observed in rat kidney lysates (calculated MW of 61.0 kDa according to NP_113721.3). Recommended concentration: 0.1-0.3 ?g/mL.

Ugt2b antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Ugt2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR