Anti-PKP2 Goat Polyclonal Antibody

Supplier: ProSci
42-422
10114-444EA 701.94 USD
10114-444
Anti-PKP2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 100 kDa band observed in mouse and rat heart lysates (calculated MW of 88.1 kDa according to mouse NP_080439.1 and 87.8 kDa according to rat NP_001093969.1). Recommended concentration: 0.3-1 ?g/mL.

PKP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PKP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR