Anti-IRF2BP1 Goat Polyclonal Antibody

Supplier: ProSci
42-415
10113-878EA 701.76 USD
10113-878
Anti-IRF2BP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 75 kDa band observed in HEPG2 lysate (calculated MW of 61.7 kDa according to NP_056464.1). Recommended concentration: 0.1-0.3 ?g/mL.

IRF2BP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IRF2BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR