Anti-FOXI3 Goat Polyclonal Antibody

Supplier: ProSci

42-411
10113-872EA 560.36 USD
10113-872
Anti-FOXI3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 37 kDa band observed in mouse and rat skin lysates (calculated MW of 42.0 kDa according to NP_001094934.1). Recommended concentration: 1-3 ?g/mL.

Foxi3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Foxi3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR