Anti-SH3GL1 Goat Polyclonal Antibody

Supplier: ProSci
42-410
10114-432EA 701.94 USD
10114-432
Anti-SH3GL1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 48 kDa band observed in K562 lysate (calculated MW of 41.5 kDa according to NP_003016.1. The observed molecular weight corresponds to earlier findings in literature with different antibodies (So et al, Proc Natl Acad Sci U S A. 1997 Mar 18;94 (6) :2563-8.; PMID: 9122235). Recommended concentration: 0.3-1 ?g/mL.

SH3GL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SH3GL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR