Anti-CRTC2 Goat Polyclonal Antibody

Supplier: ProSci
42-407
10113-868EA 701.94 USD
10113-868
Anti-CRTC2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 85 kDa band observed in Jurkat lysate (calculated MW of 73.3 kDa according to NP_859066.1). Recommended concentration: 0.3-1 ?g/mL.

CRTC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CRTC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR