Anti-ACPP Goat Polyclonal Antibody

Supplier: ProSci
42-397
10113-858EA 701.94 USD
10113-858
Anti-ACPP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 50 kDa band observed in human prostate lysates (calculated MW of 48.3 kDa according to NP_001127666.1). Recommended concentration: 0.01-0.03 ?g/mL.

ACPP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR