Anti-IDH1 Goat Polyclonal Antibody

Supplier: ProSci
42-395
10113-856EA 701.94 USD
10113-856
Anti-IDH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 38 kDa band observed in wildtype lysates of S.cerevisiae (calculated MW of 39.3 kDa according to NP_014361.1). Data kindly provided by F. Reggiiori, University Medical Centre Utrecht, Netherlands. Recommended concentration: 0.3-1 ?g/mL.

IDH1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR