Anti-IDH3G Goat Polyclonal Antibody

Supplier: ProSci
42-379
10114-402EA 701.94 USD
10114-402
Anti-IDH3G Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 38 kDa band observed in MCF7 lysate and of human, mouse and rat heart (calculated MW of 41.5 kDa according to NP_777358.1). Recommended concentration: 0.5-2 ?g/mL.

IDH3G antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IDH3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR