Anti-PRKD1 Goat Polyclonal Antibody

Supplier: ProSci
42-378
10114-400EA 617.84 USD
10114-400
Anti-PRKD1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Our routinely used Western Blotting protocol does not allow detection of proteins as large as the calculated size of 463 kDa according to NP_000287.3.

PKD1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR