Anti-PAPD5 Goat Polyclonal Antibody

Supplier: ProSci
42-370
10113-834EA 701.94 USD
10113-834
Anti-PAPD5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 75-80 kDa band observed in human and mouse brain lysates (calculated MW of 75.8 kDa according to human NP_001035374.2 and 74.2 kDa according to mouse NP_001157969.1). Recommended concentration: 0.3-1 ?g/mL.

PAPD5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PAPD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR