Anti-ITIH4 Goat Polyclonal Antibody

Supplier: ProSci

42-366
10113-826EA 560.36 USD
10113-826
Anti-ITIH4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 100 kDa band observed in rat liver lysates (calculated MW of 104 kDa according to rat NP_062242.2). Recommended concentration: 0.03-0.1 ?g/mL.

Itih4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Itih4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR